Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pax7
Entrez Gene ID
500574
Gene Name
paired box 7
Gene Aliases
RGD1564360
Chromosome Location
Chr.5: 158212922 - 158313510 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191984.1 NP_001178913.1 8 - 9 1389 81
XM_006239263.2 XP_006239325.1 8 - 9 1479 81

View Full Product Details