Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rlf
Entrez Gene ID
313566
Gene Name
rearranged L-myc fusion
Gene Aliases
-
Chromosome Location
Chr.5: 140107144 - 140162696 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_233485.8 XP_233485.6 7 - 8 1130 115

View Full Product Details