Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex10
Entrez Gene ID
680424
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
-
Chromosome Location
Chr.5: 172469978 - 172475144 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109405.1 NP_001102875.1 3 - 4 687 83

View Full Product Details