Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pla2g2c
- Entrez Gene ID
-
29387
- Gene Name
- phospholipase A2, group IIC
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 157163646 - 157187654 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U07798.1 |
- |
1 - 2 |
553 |
80 |
View Full Product Details