Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyzl4
Entrez Gene ID
363168
Gene Name
lysozyme-like 4
Gene Aliases
RGD1308401
Chromosome Location
Chr.8: 130219984 - 130228078 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246183.1 NP_001233112.1 3 - 4 365 74
XM_006244123.2 XP_006244185.2 4 - 5 1608 74
XM_006244125.2 XP_006244187.1 4 - 5 561 74
GenBank mRNA HM125534.1 - 3 - 4 365 74

View Full Product Details