Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyzl4
Entrez Gene ID
363168
Gene Name
lysozyme-like 4
Gene Aliases
RGD1308401
Chromosome Location
Chr.8: 130219984 - 130228078 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001246183.1 NP_001233112.1 2 - 3 291 73
XM_006244123.2 XP_006244185.2 3 - 4 1534 73
XM_006244125.2 XP_006244187.1 3 - 4 487 73
GenBank mRNA HM125534.1 - 2 - 3 291 73

View Full Product Details