The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031154.1 | NP_112416.1 | 6 - 7 | 478 | 83 |
GenBank mRNA | BC059130.1 | - | 6 - 7 | 536 | 83 |
FQ214048.1 | - | 6 - 7 | 527 | 83 | |
J02744.1 | - | 6 - 7 | 478 | 83 |