Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dbt
Entrez Gene ID
29611
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
-
Chromosome Location
Chr.2: 219563783 - 219592651 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053312.1 NP_445764.1 8 - 9 1034 68
GenBank mRNA BC101904.1 - 9 - 10 1246 68
BC166487.1 - 8 - 9 1021 68

View Full Product Details