Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc7
Entrez Gene ID
310787
Gene Name
fibronectin type III domain containing 7
Gene Aliases
RGD1309189
Chromosome Location
Chr.2: 211655482 - 211690799 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107717.1 NP_001101187.1 5 - 6 1164 127
XM_006233159.2 XP_006233221.1 6 - 7 1172 127
XM_006233160.2 XP_006233222.1 6 - 7 1165 127

View Full Product Details