Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifi44
Entrez Gene ID
310969
Gene Name
interferon-induced protein 44
Gene Aliases
-
Chromosome Location
Chr.2: 256897765 - 256915591 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107729.1 NP_001101199.1 6 - 7 1219 73
XM_006233495.2 XP_006233557.1 6 - 7 1247 73
XM_006233496.2 XP_006233558.1 6 - 7 1177 73
XM_006233497.2 XP_006233559.1 6 - 7 1247 73
XM_006233498.2 XP_006233560.1 6 - 7 1177 73
GenBank mRNA BC088420.1 - 7 - 8 1243 73
BC158758.1 - 6 - 7 1111 73

View Full Product Details