Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhb
Entrez Gene ID
301168
Gene Name
EF hand domain family, member B
Gene Aliases
RGD1566407
Chromosome Location
Chr.9: 47988 - 120800 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106879.1 NP_001100349.1 2 - 3 950 100
GenBank mRNA BC166482.1 - 2 - 3 951 100

View Full Product Details