Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Csn2
- Entrez Gene ID
-
29173
- Gene Name
- casein beta
- Gene Aliases
-
Csnb
- Chromosome Location
-
Chr.14: 21972274 - 21979506 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
J00711.1 |
- |
3 - 4 |
134 |
104 |
View Full Product Details