Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx25
Entrez Gene ID
58856
Gene Name
DEAD (Asp-Glu-Ala-Asp) box helicase 25
Gene Aliases
GRTH
Chromosome Location
Chr.8: 36744686 - 36760720 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031630.2 NP_113818.2 5 - 6 428 82
GenBank mRNA AF142629.2 - 5 - 6 428 82
BC078791.1 - 5 - 6 410 82

View Full Product Details