Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb1
Entrez Gene ID
316164
Gene Name
SATB homeobox 1
Gene Aliases
-
Chromosome Location
Chr.9: 2181132 - 2277949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012129.2 NP_001012129.1 3 - 4 654 128
XM_006244219.2 XP_006244281.1 3 - 4 769 128
XM_006244220.2 XP_006244282.1 3 - 4 2683 128
XM_006244221.2 XP_006244283.1 3 - 4 651 128
XM_008766760.1 XP_008764982.1 2 - 3 1314 128
XM_008766761.1 XP_008764983.1 3 - 4 1283 128
XM_008766762.1 XP_008764984.1 4 - 5 688 128
XM_008766763.1 XP_008764985.1 4 - 5 3781 128
XM_008766764.1 XP_008764986.1 2 - 3 1314 128
GenBank mRNA BC085814.1 - 3 - 4 655 128

View Full Product Details