Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc45a2
Entrez Gene ID
310152
Gene Name
solute carrier family 45, member 2
Gene Aliases
Matp
Chromosome Location
Chr.2: 60966789 - 60999398 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107653.1 NP_001101123.1 6 - 7 1481 80

View Full Product Details