Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
G6b
Entrez Gene ID
406865
Gene Name
immunoreceptor tyrosine-based inhibitory motif (ITIM) containing platelet receptor
Gene Aliases
C6orf25, Ng31
Chromosome Location
Chr.20: 5054421 - 5057026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003976.1 NP_001003976.1 3 - 4 496 140

View Full Product Details