Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdss1
Entrez Gene ID
100364990
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 1
Gene Aliases
-
Chromosome Location
Chr.17: 90033432 - 90072509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751734.3 XP_003751782.2 3 - 4 1435 110
XM_008771923.1 XP_008770145.1 3 - 4 1475 110
GenBank mRNA BC158695.1 - 3 - 4 315 110

View Full Product Details