Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dbf4
Entrez Gene ID
312046
Gene Name
DBF4 zinc finger
Gene Aliases
RGD1305854
Chromosome Location
Chr.4: 22898527 - 22914091 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191748.1 NP_001178677.1 - 363 135

Gene Symbol
LOC103690019
Entrez Gene ID
103690019
Gene Name
protein DBF4 homolog A-like
Gene Aliases
-
Chromosome Location
Chr.4: 22661991 - 22686164 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008762747.1 XP_008760969.1 2 - 3 444 135

View Full Product Details