Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prim1
Entrez Gene ID
246327
Gene Name
primase, DNA, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.7: 2435553 - 2451169 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ011608.1 - 9 - 10 986 127
BC088272.1 - 9 - 10 993 127
FQ231746.1 - 2 - 3 192 127

View Full Product Details