Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn6
Entrez Gene ID
363332
Gene Name
UBX domain protein 6
Gene Aliases
Ubxd1
Chromosome Location
Chr.9: 10995396 - 11000643 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108812.1 NP_001102282.1 4 - 5 498 75
XM_006244334.2 XP_006244396.2 5 - 6 788 75

View Full Product Details