Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plin5
Entrez Gene ID
501283
Gene Name
perilipin 5
Gene Aliases
Oxpat
Chromosome Location
Chr.9: 10956013 - 10961782 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134637.1 NP_001128109.1 5 - 6 495 136
XM_008766770.1 XP_008764992.1 5 - 6 538 136

View Full Product Details