The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014107.1 | NP_001014129.1 | 5 - 6 | 603 | 139 |
XM_006257631.2 | XP_006257693.1 | 4 - 5 | 649 | 139 | |
XM_006257632.2 | XP_006257694.1 | 5 - 6 | 688 | 139 | |
XM_006257633.2 | XP_006257695.1 | 4 - 5 | 646 | 139 | |
XM_006257634.2 | XP_006257696.1 | 4 - 5 | 646 | 139 | |
GenBank mRNA | BC086521.1 | - | 5 - 6 | 603 | 139 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006251400.2 | XP_006251462.1 | 1 - 2 | 621 | 139 |
XM_008770405.1 | XP_008768627.1 | 1 - 2 | 621 | 139 |