Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cops6
Entrez Gene ID
304343
Gene Name
COP9 signalosome subunit 6
Gene Aliases
-
Chromosome Location
Chr.12: 19303387 - 19306226 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107129.1 NP_001100599.1 3 - 4 381 94

View Full Product Details