The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133538.2 | NP_598222.2 | 1 - 2 | 47 | 75 |
GenBank mRNA | AF448818.1 | - | 1 - 2 | 47 | 75 |