The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108121.1 | NP_001101591.1 | 1 - 2 | 419 | 77 |
XM_006242474.2 | XP_006242536.1 | 2 - 3 | 518 | 77 | |
XM_006242476.1 | XP_006242538.1 | 2 - 3 | 407 | 77 | |
XM_006242477.2 | XP_006242539.1 | 2 - 3 | 266 | 77 |