Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps28
Entrez Gene ID
691531
Gene Name
ribosomal protein S28
Gene Aliases
-
Chromosome Location
Chr.7: 18682071 - 18683440 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105730.1 NP_001099200.1 2 - 3 113 131
GenBank mRNA FQ210333.1 - 2 - 3 105 131
FQ211223.1 - 2 - 3 105 131
FQ217403.1 - 2 - 3 110 136
FQ218290.1 - 2 - 3 104 131
FQ221642.1 - 2 - 3 105 131
FQ221735.1 - 2 - 3 106 131
FQ222008.1 - 2 - 3 105 131
FQ222172.1 - 2 - 3 105 131
FQ222320.1 - 2 - 3 105 131
FQ222499.1 - 2 - 3 105 131
FQ222594.1 - 2 - 3 108 131
FQ222598.1 - 2 - 3 106 131
FQ222664.1 - 2 - 3 105 131
FQ223680.1 - 2 - 3 106 131
FQ224357.1 - 2 - 3 105 131
FQ228386.1 - 2 - 3 106 131
FQ228826.1 - 2 - 3 104 131
FQ228989.1 - 2 - 3 105 131
FQ229601.1 - 2 - 3 105 131
X59277.1 - 2 - 3 94 131

View Full Product Details