The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201420.2 | NP_958823.2 | 6 - 7 | 1276 | 60 |
XM_006235691.2 | XP_006235753.1 | 6 - 7 | 1245 | 60 | |
GenBank mRNA | AY510602.1 | - | 6 - 7 | 1067 | 60 |
BC097260.1 | - | 6 - 7 | 1276 | 60 |