Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab22a
Entrez Gene ID
366265
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 171743774 - 171790558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235705.2 XP_006235767.1 3 - 4 592 158
XM_008762529.1 XP_008760751.1 3 - 4 592 158
GenBank mRNA BC158650.1 - 3 - 4 416 158

View Full Product Details