Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd29
Entrez Gene ID
498823
Gene Name
ankyrin repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.18: 3673915 - 3676187 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190372.1 NP_001177301.1 - 724 93

View Full Product Details