Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hfe
Entrez Gene ID
29199
Gene Name
hemochromatosis
Gene Aliases
-
Chromosome Location
Chr.17: 43661276 - 43669327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173434.1 NP_001166905.1 1 - 2 173 104
GenBank mRNA AF176534.1 - 1 - 2 93 104
FQ219597.1 - 1 - 2 187 104

View Full Product Details