Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmnn
Entrez Gene ID
291137
Gene Name
geminin
Gene Aliases
-
Chromosome Location
Chr.17: 42302527 - 42310783 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253927.2 XP_006253989.1 1 - 2 130 131

View Full Product Details