The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007619.1 | NP_001007620.1 | 3 - 4 | 546 | 125 |
GenBank mRNA | AY539871.1 | - | - | 458 | 125 |
BC079452.1 | - | 3 - 4 | 546 | 125 |