Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcgf3
Entrez Gene ID
305624
Gene Name
polycomb group ring finger 3
Gene Aliases
Rnf3
Chromosome Location
Chr.14: 2247805 - 2297155 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107245.2 NP_001100715.2 2 - 3 158 62
GenBank mRNA BC167000.1 - 2 - 3 158 62

View Full Product Details