Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mt4
Entrez Gene ID
498911
Gene Name
metallothionein 4
Gene Aliases
RGD1565500
Chromosome Location
Chr.19: 11339767 - 11341863 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126084.1 NP_001119556.1 2 - 3 126 117

View Full Product Details