The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008324.1 | NP_001008325.1 | 14 - 15 | 1769 | 102 |
GenBank mRNA | BC085933.1 | - | 14 - 15 | 1769 | 102 |