Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spryd3
Entrez Gene ID
315327
Gene Name
SPRY domain containing 3
Gene Aliases
RGD1561500
Chromosome Location
Chr.7: 143721088 - 143738237 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191790.1 NP_001178719.1 8 - 9 901 73

View Full Product Details