Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Golph3l
Entrez Gene ID
310669
Gene Name
golgi phosphoprotein 3-like
Gene Aliases
-
Chromosome Location
Chr.2: 197720259 - 197749060 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007698.1 NP_001007699.1 3 - 4 467 62
GenBank mRNA BC081987.1 - 3 - 4 467 62

View Full Product Details