Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekho1
Entrez Gene ID
310674
Gene Name
pleckstrin homology domain containing, family O member 1
Gene Aliases
Ckip1, RGD1311487
Chromosome Location
Chr.2: 198112104 - 198120202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025119.1 NP_001020290.1 4 - 5 559 73
GenBank mRNA BC091420.1 - 4 - 5 559 73
FQ230444.1 - 3 - 4 368 73
FQ230757.1 - 3 - 4 409 73
FQ232601.1 - 2 - 3 172 73

View Full Product Details