Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vangl1
Entrez Gene ID
690366
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 204575596 - 204625835 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109584.1 NP_001103054.1 6 - 7 1296 152
XM_006233054.2 XP_006233116.1 7 - 8 1376 152
XM_006233055.2 XP_006233117.1 7 - 8 1339 152

View Full Product Details