The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011951.1 | NP_001011951.1 | 2 - 3 | 401 | 123 |
GenBank mRNA | BC078997.1 | - | 2 - 3 | 401 | 123 |