Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hist2h2be
Entrez Gene ID
295274
Gene Name
histone cluster 2, H2be
Gene Aliases
RGD1562346
Chromosome Location
Chr.2: 198360810 - 198363399 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_227459.5 XP_227459.1 1 - 1 2232 85

View Full Product Details