Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fmo5
Entrez Gene ID
246248
Gene Name
flavin containing monooxygenase 5
Gene Aliases
-
Chromosome Location
Chr.2: 199796870 - 199823927 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144739.1 NP_653340.1 1 - 2 303 91
GenBank mRNA AF458413.1 - 1 - 2 303 91

View Full Product Details