The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022589.1 | NP_072111.1 | 5 - 6 | 470 | 106 |
GenBank mRNA | AJ271442.1 | - | 5 - 6 | 470 | 106 |
BC085733.1 | - | 5 - 6 | 490 | 106 |