Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim45
Entrez Gene ID
295323
Gene Name
tripartite motif-containing 45
Gene Aliases
-
Chromosome Location
Chr.2: 203301838 - 203310532 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106453.1 NP_001099923.1 5 - 6 2035 119

View Full Product Details