Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctsb
Entrez Gene ID
64529
Gene Name
cathepsin B
Gene Aliases
-
Chromosome Location
Chr.15: 46316741 - 46337613 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022597.2 NP_072119.2 3 - 4 348 137
GenBank mRNA BC072490.1 - 3 - 4 311 137
X82396.1 - 3 - 4 277 137

View Full Product Details