Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt7
Entrez Gene ID
29750
Gene Name
polypeptide N-acetylgalactosaminyltransferase 7
Gene Aliases
-
Chromosome Location
Chr.16: 35935035 - 36062734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022926.1 NP_075215.1 5 - 6 1014 122
GenBank mRNA AF076167.1 - 5 - 6 1014 122

View Full Product Details