Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stk11
Entrez Gene ID
314621
Gene Name
serine/threonine kinase 11
Gene Aliases
Lkb1
Chromosome Location
Chr.7: 12440751 - 12457513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108069.1 NP_001101539.1 9 - 10 2149 80

View Full Product Details