Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vsx1
Entrez Gene ID
689704
Gene Name
visual system homeobox 1
Gene Aliases
-
Chromosome Location
Chr.3: 146484238 - 146491837 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109546.1 NP_001103016.1 2 - 3 694 73

View Full Product Details