Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dusp15
Entrez Gene ID
362238
Gene Name
dual specificity phosphatase 15
Gene Aliases
-
Chromosome Location
Chr.3: 148417990 - 148428494 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108598.2 NP_001102068.2 5 - 6 466 115
GenBank mRNA CB576376.1 - 5 - 6 343 115

View Full Product Details