Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Herpud1
Entrez Gene ID
85430
Gene Name
homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1
Gene Aliases
Sup
Chromosome Location
Chr.19: 11046903 - 11057570 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053523.1 NP_445975.1 3 - 4 398 128
GenBank mRNA AB033771.1 - 3 - 4 398 128
BC061849.1 - 3 - 4 420 128

View Full Product Details